4-Bromo-5-fluoro-2-methylphenyl methyl ether ≥97%

Supplier: AOBChem USA

Warning

45369-1G 45369-25G 45369-500MG 45369-5G
76889-932EA 393.6 USD
76889-932 76889-934 76889-936 76889-938
4-Bromo-5-fluoro-2-methylphenyl methyl ether ≥97%
4-Bromo-5-fluoro-2-methylphenyl methyl ether
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD11840965
CAS Number: 901231-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR