[6-(2-Methoxyethoxy)-5-methylpyridin-3-yl]boronic acid ≥97%

Supplier: AOBChem USA

Warning

42514-1G 42514-25G 42514-500MG 42514-5G
76889-874EA 292.81 USD
76889-874 76889-876 76889-878 76889-880
[6-(2-Methoxyethoxy)-5-methylpyridin-3-yl]boronic acid ≥97%
[6-(2-Methoxyethoxy)-5-methylpyridin-3-yl]boronic acid
Formula: C₉H₁₄BNO₄
MW: 211.023 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27935552
CAS Number: 2377608-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR