4-Fluoro-2-nitrobenzeneboronic acidpinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

41214-1G 41214-25G 41214-500MG 41214-5G
76889-848EA 155.96 USD
76889-848 76889-850 76889-852 76889-854
4-Fluoro-2-nitrobenzeneboronic acidpinacol ester ≥97%
4-Fluoro-2-nitrobenzeneboronic acidpinacol ester
Formula: C₁₂H₁₅BFNO₄
MW: 267.063 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996295
CAS Number: 1288978-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR