4'-Methoxybiphenyl-4-ylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-[4-(4-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
39608-1G
39608-25G
39608-5G
76889-768EA
79.57
USD
76889-768
76889-770
76889-772
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester ≥97%
4'-Methoxybiphenyl-4-ylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₉H₂₃BO₃ MW: 310.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294530 CAS Number: 446311-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...