1-Bromo-4-fluoro-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

38840-1G 38840-25G 38840-500MG 38840-5G
76889-752EA 173.99 USD
76889-752 76889-754 76889-756 76889-758
1-Bromo-4-fluoro-2-methoxy-5-methylbenzene ≥97%
1-Bromo-4-fluoro-2-methoxy-5-methylbenzene
Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD15527526
CAS Number: 314298-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR