1,5-Dibromo-2-methoxy-3-(trifluoromethyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

38816-1G 38816-25G 38816-500MG 38816-5G
76889-730EA 173.99 USD
76889-730 76889-732 76889-734 76889-736
1,5-Dibromo-2-methoxy-3-(trifluoromethyl)benzene ≥97% research grade
1,5-Dibromo-2-methoxy-3-(trifluoromethyl)benzene
Formula: C₈H₅Br₂F₃O
MW: 333.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31617585
CAS Number: 1124144-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR