1-Chloro-2-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

38721-1G 38721-25G 38721-500MG 38721-5G
76889-714EA 144.29 USD
76889-714 76889-716 76889-718 76889-720
1-Chloro-2-ethoxy-3-fluorobenzene ≥97%
1-Chloro-2-ethoxy-3-fluorobenzene
Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28126791
CAS Number: 1668474-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR