1-Bromo-2-chloro-4-fluoro-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

38697-1G 38697-25G 38697-500MG 38697-5G
76889-698EA 381.93 USD
76889-698 76889-700 76889-702 76889-704
1-Bromo-2-chloro-4-fluoro-3-methoxybenzene ≥95%
1-Bromo-2-chloro-4-fluoro-3-methoxybenzene
Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28402076
CAS Number: 943830-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR