2-(4-Methoxyphenyl)-1,3,4-oxadiazole ≥97%

Supplier: AOBChem USA

Warning

38539-1G 38539-25G 38539-500MG 38539-5G
76889-628EA 206.88 USD
76889-628 76889-630 76889-632 76889-634
2-(4-Methoxyphenyl)-1,3,4-oxadiazole ≥97%
2-(4-Methoxyphenyl)-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00491615
CAS Number: 829-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR