(5-Chloro-2-(trifluoromethyl)phenyl)(morpholino)methanone ≥97% research grade

Supplier: AOBChem USA

Warning

38489-1G 38489-25G 38489-5G 38489-500MG
76889-604EA 106.09 USD
76889-604 76889-606 76889-608 77652-750
(5-Chloro-2-(trifluoromethyl)phenyl)(morpholino)methanone ≥97% research grade
(5-Chloro-2-(trifluoromethyl)phenyl)(morpholino)methanone
Formula: C₁₂H₁₁ClF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32707087
CAS Number: 2413441-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR