(5-Chloro-2-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

38486-1G 38486-25G 38486-5G 38486-500MG
76889-592EA 94.43 USD
76889-592 76889-594 76889-596 77646-566
(5-Chloro-2-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(5-Chloro-2-(trifluoromethyl)phenyl)(4-methylpiperazin-1-yl)methanone
Formula: C₁₃H₁₄ClF₃N₂O
MW: 306.711 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707084
CAS Number: 2413441-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR