2-(Pyridin-3-yloxy)ethanol ≥97%

Supplier: AOBChem USA

Warning

38481-1G 38481-25G 38481-500MG 38481-5G
76889-578EA 231.28 USD
76889-578 76889-580 76889-582 76889-584
2-(Pyridin-3-yloxy)ethanol ≥97%
2-(Pyridin-3-yloxy)ethanol
Formula: C₇H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13248572
CAS Number: 119967-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR