(E)-3-(3,5-dichloro-4-methoxyphenyl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

38341-1G 38341-25G 38341-500MG 38341-5G
76889-512EA 290.69 USD
76889-512 76889-514 76889-516 76889-518
(E)-3-(3,5-dichloro-4-methoxyphenyl)acrylic acid ≥97%
(E)-3-(3,5-dichloro-4-methoxyphenyl)acrylic acid
Formula: C₁₀H₈Cl₂O₃
MW: 247.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09999375
CAS Number: 24076-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR