1-(3-Iodophenyl)-2,5-dimethylpyrrole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Iodophenyl)-2,5-dimethyl-1H-pyrrole

38282-1G 38282-25G 38282-5G
76889-488EA 66.84 USD
76889-488 76889-490 76889-492
1-(3-Iodophenyl)-2,5-dimethylpyrrole ≥97%
1-(3-Iodophenyl)-2,5-dimethylpyrrole

Precaución: for Research Use Only

Formula: C₁₂H₁₂IN
Storage Temperature: Ambient
MDL Number: MFCD07429165
CAS Number: 217314-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR