1-Chloro-4-(propan-2-yloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-4-isopropoxybenzene

38274-1G 38274-25G 38274-500MG 38274-5G
76889-474EA 183.54 USD
76889-474 76889-476 76889-478 76889-480
1-Chloro-4-(propan-2-yloxy)benzene ≥97%
1-Chloro-4-(propan-2-yloxy)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MW: 170.636 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070779
CAS Number: 51241-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR