(3,5-Difluoro-4-propoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

38263-1G 38263-25G 38263-5G
76889-460EA 96.55 USD
76889-460 76889-462 76889-464
(3,5-Difluoro-4-propoxyphenyl)boronic acid ≥97%
(3,5-Difluoro-4-propoxyphenyl)boronic acid
Formula: C₉H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20231441
CAS Number: 2096331-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR