(E)-ethyl 3-(3-chloro-4-methoxyphenyl)acrylate ≥97%

Supplier: AOBChem USA

Warning

38113-1G 38113-500MG 38113-5G
76889-332EA 473.17 USD
76889-332 76889-334 76889-336
(E)-ethyl 3-(3-chloro-4-methoxyphenyl)acrylate ≥97%
(E)-ethyl 3-(3-chloro-4-methoxyphenyl)acrylate
Formula: C₁₂H₁₃ClO₃
MW: 240.683 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20440907
CAS Number: 17384-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR