2-Bromo-4,5-dichloroanisole ≥97%

Supplier: AOBChem USA

Warning

38091-1G 38091-25G 38091-500MG 38091-5G
76889-302EA 137.92 USD
76889-302 76889-304 76889-306 76889-308
2-Bromo-4,5-dichloroanisole ≥97%
2-Bromo-4,5-dichloroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD11845923
CAS Number: 174913-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR