1-Bromo-3,4-difluoro-2,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

38079-1G 38079-500MG 38079-5G
76889-292EA 589.87 USD
76889-292 76889-294 76889-296
1-Bromo-3,4-difluoro-2,5-dimethoxybenzene ≥97%
1-Bromo-3,4-difluoro-2,5-dimethoxybenzene
Formula: C₈H₇BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32707052
CAS Number: 2432848-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR