(3-Bromo-5-nitrophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

38066-25G 38066-5G
76889-282EA 488.02 USD
76889-282 76889-284
(3-Bromo-5-nitrophenyl)(thiomorpholino)methanone ≥97%
(3-Bromo-5-nitrophenyl)(thiomorpholino)methanone
Formula: C₁₁H₁₁BrN₂O₃S
MW: 331.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707051
CAS Number: 2432848-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR