1-(3-Bromo-5-nitrobenzoyl)piperidine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-nitrophenyl)(piperidin-1-yl)methanone

38064-25G 38064-5G
76889-274EA 595.17 USD
76889-274 76889-276
1-(3-Bromo-5-nitrobenzoyl)piperidine ≥97% research grade
1-(3-Bromo-5-nitrobenzoyl)piperidine
Formula: C₁₂H₁₃BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09475858
CAS Number: 941294-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR