6-Chloro-2-fluoro-3-methylanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-3-fluoro-2-methoxy-4-methylbenzene

38019-25G 38019-5G
76889-262EA 418 USD
76889-262 76889-264
6-Chloro-2-fluoro-3-methylanisole ≥97%
6-Chloro-2-fluoro-3-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631377
CAS Number: 261762-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR