(3-Bromo-5-chlorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

37944-1G 37944-25G 37944-500MG 37944-5G
76889-218EA 441.34 USD
76889-218 76889-220 76889-222 76889-224
(3-Bromo-5-chlorophenyl)(morpholino)methanone ≥97%
(3-Bromo-5-chlorophenyl)(morpholino)methanone
Formula: C₁₁H₁₁BrClNO₂
MW: 304.568 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31613939
CAS Number: 1327122-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR