2-(Benzyloxy)-4-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

37924-1G 37924-25G 37924-500MG 37924-5G
76889-202EA 260.99 USD
76889-202 76889-204 76889-206 76889-208
2-(Benzyloxy)-4-(trifluoromethyl)benzoic acid ≥97% research grade
2-(Benzyloxy)-4-(trifluoromethyl)benzoic acid
Formula: C₁₅H₁₁F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD27922761
CAS Number: 1822843-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR