1-(3-Bromophenyl)pyrrole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Bromophenyl)-1H-pyrrole, N-(3-Bromophenyl)pyrrole

37920-1G 37920-25G 37920-5G
76889-190EA 88.06 USD
76889-190 76889-192 76889-194
1-(3-Bromophenyl)pyrrole ≥97%
1-(3-Bromophenyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02665243
CAS Number: 107302-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR