(2,5-Difluoro-4-propoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

37859-1G 37859-500MG 37859-5G 37859-25G
76889-176EA 706.56 USD
76889-176 76889-178 76889-180 77659-056
(2,5-Difluoro-4-propoxyphenyl)boronic acid ≥97%
(2,5-Difluoro-4-propoxyphenyl)boronic acid
Formula: C₉H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20529441
CAS Number: 2096331-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR