1-Bromo-4-(difluoromethoxy)-2,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

37843-1G 37843-500MG 37843-5G
76889-164EA 528.33 USD
76889-164 76889-166 76889-168
1-Bromo-4-(difluoromethoxy)-2,5-difluorobenzene ≥97%
1-Bromo-4-(difluoromethoxy)-2,5-difluorobenzene
Formula: C₇H₃BrF₄O
MW: 258.996 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28402353
CAS Number: 1394130-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR