2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2,3-Difluoro-5-methoxy-4-methylphenyl)methanol

37736-1G 37736-500MG 37736-5G
76889-156EA 706.56 USD
76889-156 76889-158 76889-160
2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol ≥95%
2,3-Difluoro-5-methoxy-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27986850
CAS Number: 1706461-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR