(E)-3-(3-bromo-2,5-dichlorophenyl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

37628-1G 37628-500MG 37628-5G
76889-116EA 599.41 USD
76889-116 76889-118 76889-120
(E)-3-(3-bromo-2,5-dichlorophenyl)acrylic acid ≥97%
(E)-3-(3-bromo-2,5-dichlorophenyl)acrylic acid
Formula: C₉H₅BrCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632247
CAS Number: 2169861-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR