(E)-3-(4-bromo-3-chlorophenyl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

37626-1G 37626-25G 37626-500MG 37626-5G
76889-100EA 256.74 USD
76889-100 76889-102 76889-104 76889-106
(E)-3-(4-bromo-3-chlorophenyl)acrylic acid ≥97%
(E)-3-(4-bromo-3-chlorophenyl)acrylic acid
Formula: C₉H₆BrClO₂
MW: 261.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06358688
CAS Number: 790681-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR