1-Methyl-1H-indol-5-ol ≥97%

Supplier: AOBChem USA

Warning

37620-1G 37620-500MG 37620-5G 37620-25G
76889-088EA 530.45 USD
76889-088 76889-090 76889-092 77659-042
1-Methyl-1H-indol-5-ol ≥97%
1-Methyl-1H-indol-5-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12924270
CAS Number: 13523-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR