2,3-Dichloro-6-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,3-Dichloro-6-fluorophenyl)methanol

37577-25G 37577-5G 37577-1G 37577-500MG
76889-080EA 595.17 USD
76889-080 76889-082 77642-470 77650-480
2,3-Dichloro-6-fluorobenzyl alcohol ≥97%
2,3-Dichloro-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD04115872
CAS Number: 886497-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR