2-(2-Fluoro-6-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

37540-25G 37540-5G
76889-072EA 537.88 USD
76889-072 76889-074
2-(2-Fluoro-6-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-6-(trifluoromethyl)phenyl)-1,3-dioxolane
Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209159
CAS Number: 773102-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR