(2-Fluoro-5-methylpyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

37522-1G 37522-25G 37522-500MG 37522-5G
76889-056EA 350.1 USD
76889-056 76889-058 76889-060 76889-062
(2-Fluoro-5-methylpyridin-3-yl)(pyrrolidin-1-yl)methanone ≥97%
(2-Fluoro-5-methylpyridin-3-yl)(pyrrolidin-1-yl)methanone
Formula: C₁₁H₁₃FN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD32632228
CAS Number: 2404734-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR