1-Bromo-4-iodo-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

37510-1G 37510-25G 37510-500MG 37510-5G
76889-042EA 432.85 USD
76889-042 76889-044 76889-046 76889-048
1-Bromo-4-iodo-2-isopropylbenzene ≥95%
1-Bromo-4-iodo-2-isopropylbenzene
Formula: C₉H₁₀BrI
Storage Temperature: Refrigerator
MDL Number: MFCD29094024
CAS Number: 1369927-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR