2,3-Difluoro-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37483-1G 37483-25G 37483-500MG 37483-5G
76889-028EA 172.93 USD
76889-028 76889-030 76889-032 76889-034
2,3-Difluoro-4-isopropoxybenzoic acid ≥97%
2,3-Difluoro-4-isopropoxybenzoic acid
Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632225
CAS Number: 2404733-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR