1-Chloro-2,3-difluoro-4-methoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

37469-1G 37469-25G 37469-500MG 37469-5G
76889-016EA 379.81 USD
76889-016 76889-018 76889-020 76889-022
1-Chloro-2,3-difluoro-4-methoxybenzene ≥97% research grade
1-Chloro-2,3-difluoro-4-methoxybenzene
Formula: C₇H₅ClF₂O
MW: 178.564 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415548
CAS Number: 1261808-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR