(4-Ethoxy-2,3-Difluorophenyl)(Methyl)Sulfane ≥95% research grade

Supplier: AOBChem USA

Warning

37459-25G 37459-5G
76889-004EA 453.01 USD
76889-004 76889-006
(4-Ethoxy-2,3-Difluorophenyl)(Methyl)Sulfane ≥95% research grade
(4-Ethoxy-2,3-Difluorophenyl)(Methyl)Sulfane
Formula: C₉H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD32707027
CAS Number: 2484889-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR