2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
37401-1G
37401-25G
37401-5G
76888-990EA
62.6
USD
76888-990
76888-992
76888-994
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine ≥97%
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₁₄H₁₁BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00444784 CAS Number: 1023-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...