1,5-Dibromo-2-ethoxy-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

37332-1G 37332-25G 37332-5G
76888-978EA 62.6 USD
76888-978 76888-980 76888-982
1,5-Dibromo-2-ethoxy-3-fluorobenzene ≥97%
1,5-Dibromo-2-ethoxy-3-fluorobenzene
Formula: C₈H₇Br₂FO
MW: 297.947 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784533
CAS Number: 1803776-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR