4-Fluoro-3-methylbenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

37313-25G 37313-5G
76888-968EA 198.39 USD
76888-968 76888-970
4-Fluoro-3-methylbenzeneboronic acid pinacol ester ≥97%
4-Fluoro-3-methylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₈BFO₂
MW: 236.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729904
CAS Number: 1001200-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR