1-(4-Chlorophenyl)pyrrole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Chlorophenyl)-1H-pyrrole, N-(4-Chlorophenyl)pyrrole

37273-25G 37273-5G
76888-946EA 488.02 USD
76888-946 76888-948
1-(4-Chlorophenyl)pyrrole ≥97%
1-(4-Chlorophenyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047067
CAS Number: 5044-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR