(2-Chloro-6-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97% research grade

Supplier: AOBChem USA

Warning

37203-1G 37203-25G 37203-500MG 37203-5G
76888-890EA 140.04 USD
76888-890 76888-892 76888-894 76888-896
(2-Chloro-6-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97% research grade
(2-Chloro-6-(trifluoromethyl)phenyl)(thiomorpholino)methanone
Formula: C₁₂H₁₁ClF₃NOS
Storage Temperature: Refrigerator
MDL Number: MFCD32632160
CAS Number: 2379321-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR