2-(m-Tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

37106-1G 37106-25G 37106-500MG 37106-5G
76888-862EA 201.58 USD
76888-862 76888-864 76888-866 76888-868
2-(m-Tolyl)pyridine ≥97%
2-(m-Tolyl)pyridine
Formula: C₁₂H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD06201378
CAS Number: 4373-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR