1-(Difluoromethoxy)-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

37023-1G 37023-25G 37023-500MG 37023-5G
76888-822EA 203.7 USD
76888-822 76888-824 76888-826 76888-828
1-(Difluoromethoxy)-2-fluoro-3-methylbenzene ≥97%
1-(Difluoromethoxy)-2-fluoro-3-methylbenzene
Formula: C₈H₇F₃O
MW: 176.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698459
CAS Number: 90370-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR