(6-Methoxy-pyridin-2-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

37007-1G 37007-25G 37007-5G
76888-810EA 94.43 USD
76888-810 76888-812 76888-814
(6-Methoxy-pyridin-2-yl)methanol ≥97% research grade
(6-Methoxy-pyridin-2-yl)methanol
Formula: C₇H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08235128
CAS Number: 63071-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR