1-(4-Chloro-3-hydroxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

36928-1G 36928-25G 36928-500MG 36928-5G
76888-730EA 281.14 USD
76888-730 76888-732 76888-734 76888-736
1-(4-Chloro-3-hydroxyphenyl)ethanone ≥97%
1-(4-Chloro-3-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.593 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00034670
CAS Number: 61124-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR