1-Methylindazole-5-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Methyl-1H-indazole-5-carboxylic acid

36894-25G 36894-5G
76888-710EA 360.71 USD
76888-710 76888-712
1-Methylindazole-5-carboxylic acid ≥97% research grade
1-Methylindazole-5-carboxylic acid
Formula: C₉H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11901714
CAS Number: 1176754-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR