2-Bromo-5-fluoro-4-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

36817-1G 36817-25G 36817-500MG 36817-5G
76888-670EA 153.84 USD
76888-670 76888-672 76888-674 76888-676
2-Bromo-5-fluoro-4-iodotoluene ≥97%
2-Bromo-5-fluoro-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Refrigerator
MDL Number: MFCD28040490
CAS Number: 1805937-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR