2-(Methoxymethyl)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36811-1G 36811-25G 36811-500MG 36811-5G
76888-654EA 225.98 USD
76888-654 76888-656 76888-658 76888-660
2-(Methoxymethyl)-4-(trifluoromethyl)benzaldehyde ≥97%
2-(Methoxymethyl)-4-(trifluoromethyl)benzaldehyde
Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632087
CAS Number: 2379322-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR